Learn More
Eppendorf™ Polycarbonate Cover Cryotube Rotor
Brand: Eppendorf™ 5427705007
478.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Cryotube Rotor | |
18 x 1.0/2 mL | |
8,320 x g | |
8,900 rpm |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Eppendorf™ Polycarbonate Cover Cryotube Rotor > Capacity: 18 x 1.0/2mL
Spot an opportunity for improvement?Share a Content Correction