missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Polycarbonate Desiccators

Product Code. 10202081 Shop All Brand Products
Click to view available options
Height (Metric):
195 mm
240 mm
300 mm
Diameter (Metric) Outer:
170 mm
235 mm
285 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10202081

Brand: Brand™ 65810

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Includes polycarbonate lid, polypropylene base and desiccant tray. BRAND™ Polycarbonate Desiccators feature a venting stopper with non-return valve in lid and polychlorobutadiene rubber (CK) O-ring compressed by vacuum to provide the seal between lid and base. Light-weight and easy to handle.

  • Includes polycarbonate lid, polypropylene base and desiccant tray.
  • Venting stopper with non-return valve in lid.
  • Polychlorobutadiene rubber (CK) O-ring, compressed by vacuum provides seal between lid and base.
  • Light-weight and easy to handle.
TRUSTED_SUSTAINABILITY

Specifications

Material Polycarbonate Lid, Polypropylene Base and Dessicant Tray
Product Type Desiccator
Includes Polychlorobutadiene rubber (CK) O-ring
Height (Metric) 240 mm
Diameter (Metric) Outer 235 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.