Learn More
Bollé Safety Polycarbonate Goggles Product Code.: 10480175
In Stock
to see stock.
Add to basket
Bollé Safety Polycarbonate Goggles
Product Code. 10480175
EN 166 1B3459 CE
Click to view available options
Certifications/Compliance:
EN 166 1B3459 CE
EN 170
Product Code. 10480175
Brand: Bollé Safety BLAPSI
In Stock
to see stock.
Edge
Add to basket
Add to basket
Let Bolle Protection™ Polycarbonate Goggles provide the affordability and flexibility that give you comfortable protection. The goggles have a wrap-around fit.
  • Clear
  • Light weight, wrap around safety spectacles
  • Polycarbonate clear anti mist and anti scratch coatings
  • Wide field of vision
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 166 1B3459 CE
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Product Type Safety Goggles
For Use With (Equipment) Protection Against Impact, Chemicals, Dust, Gas and Molten Metal
For Use With (Application) Protection Against Impact, Chemicals, Dust, Gas and Molten Metal
Frame Color Clear
No. per Pack 1 Ea.
Ventilation Details Yes
Material Polycarbonate
Product Information
  • Multiple Manufacturers
    Use code 24095 to receive this promotion.
    Promo Code: 24095
    Expires2025-04-15
    Days
    10
    Hours
    15
    Minutes
    47
    Seconds
    5
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 17/03/2025 and 15/04/2025 subject to availability of the product in stock. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy or if the product is no longer available in stock. Other restrictions may apply.
    Terms & Conditions Learn More
Product Content Correction
Product Title
Bollé Safety Polycarbonate Goggles >