Learn More
Bollé Safety Polycarbonate Goggles Product Code.: 11736575
Estimated Shipment: 08-08-2025
to see stock.
Add to basket
Bollé Safety Polycarbonate Goggles
Product Code. 11736575
EN 170
Click to view available options
Certifications/Compliance:
EN 166 1B3459 CE
EN 170
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 11736575
Brand: Bollé Safety BLEPSI
Estimated Shipment: 08-08-2025
to see stock.
Add to basket
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Let Bolle Protection™ Polycarbonate Goggles provide the affordability and flexibility that give you comfortable protection. The goggles have a wrap-around fit.
  • Clear
  • Light weight, wrap around safety spectacles
  • Polycarbonate clear anti mist and anti scratch coatings
  • Wide field of vision
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 170
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Product Type Safety Goggles
Frame Color Clear
No. per Pack 1 Ea.
Material Polycarbonate
Product Content Correction
Product Title
Bollé Safety Polycarbonate Goggles >