missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Polycarbonate/PTFE Coated Rotor

For use with Eppendorf Centrifuges

Brand:  Eppendorf™ 5427712003

Product Code. 10483232

  • £470.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Polycarbonate/PTFE Coated Rotor
  • Includes lid
  • Max. speed: 20,800 × g (14,000rpm)
  • Max. capacity: 30 × 1.5/2.0mL tubes
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Polycarbonate, PTFE
30 x 1.5/2 mL
14,000 rpm
Rotor
20,817 x g
With lid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers