Learn More
Cole-Parmer™ Polycarbonate Rotating Luer Fitting, male x male
Product Code. 11773924
Click to view available options
:
Cole-Parmer Polycarbonate Rotating Luer Fitting, male x male
Product Code. 11773924
Brand: Cole-Parmer™ 0646490
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Easily complete your luer fittings systems with these premium connectors. Cole-Parmer™ Polycarbonate Rotating Luer Fittings are ideal for many applications with small tubing.
  • Specialty plastic luer fitting
  • Standard connection for syringes
  • Rotating connection, male luer lock x male luer lock
  • Non-sterile
  • Plugs and caps are available
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Specifications
Material Polycarbonate
For Use With Syringes and many applications with small tubing.
Packaging Quantity 1 Each