missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ 4 Side Locks for Kartell™ Safety Box

Product Code. 10540243
Click to view available options
Material:
Polycarbonate
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10540243

Brand: Kartell™ 0856900

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4 safety locks preventing accidental opening if the Safety Box is dropped.

  • For use with Kartell™ Safety Box

Specifications

Color Clear
Material Polycarbonate
For Use With (Application) Transporting Tubes, Containers, Vials, Plastic Bags, Ready-to-Use Syringe
For Use With (Equipment) Kartell™ Safety Box (0056900)
Product Type Side Locks for Safety Box
Quantity Pack of 4
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.