Learn More
Masterflex™ Polycarbonate Smart Site Luer Fitting Connection
Connect these plastic luer assemblies to bio-bags or a sampling tube. Cole-Parmer™ Polycarbonate Smart Site Luer Fitting Connections are molded onto the tubing for a seamless transition from source to system.
Brand: Masterflex™ 30526-10
Code : A0
Additional Details : Weight : 0.04000kg
Description
- Each assembly comes packaged individually and can be sterilized
- Complete your luer fittings systems with these premium connectors
- Included are some special configurations available only in polycarbonate
- All female ports may be used with either slip or luer lock connections
- Plugs and caps are also available
Specifications
Smart Site Luer Fitting Connection | |
Polycarbonate | |
10 Each | |
Female x Male Luer Lock |