Learn More
Cole-Parmer™ Polycarbonate Tensioning Tools
Adjust tension from light to very tight with this tool. Cole-Parmer™ Polycarbonate Tensioning Tool features dial-adjustable tension setting. Simply squeeze the handle to pull tie to a preset tension and cut.
Brand:  Cole-Parmer™ 06830-07
Product Code. 11785049
  • £86.10 / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Shock-resistant tool absorbs all impact. Ideal for 18 to 175 lb. cable or zip ties. No twisting necessary.
Specifications
Specifications
Tensioning Tools
Uniform clamping or work in series, equipped with a cutting device
−40° to +85°C
Polycarbonate
1 Ea.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products