Learn More
Spex CertiPrep™ Polycarbonate vials
Polycarbonate cryovial with screw cap holds grinding media up to 1/2 inch and fits SPEX SamplePrep Homogeniser
Brand:  Spex CertiPrep™ 2253-PC-48
Product Code. 12839055
  • £339.00 / Pack of 48
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Polycarbonate cryovial with screw-on cap, 290mm x 114mm, 50mL. Holds grinding media up to 1/2 in and fits SPEX SamplePrep homogeniser 2196-16-PE holder and 2664 cryo-block. Grinding load per vial 20mL. Supplied as a pack of 48 vials with 48 caps.
  • Holds grinding load of 20mL per vial
  • Polycarbonate so suitable for cryogenic grinding
Specifications
Specifications
Polycarbonate
with screw-on cap
48 Pk.
50 mL
Screw Cap
Homogenizer