Learn More
Polydimethylsiloxane (silicone) emulsion, Thermo Scientific Chemicals
Description
Polydimethylsiloxane (silicone) emulsion is the most widely used silicon-based organic polymer and is particularly known for its unusual rheological (or flow) properties. Further, it is used in contact lenses, medical devices to elastomers and in shampoos (as dimethicone makes hair shiny and slippery). It finds application as an antifoaming agent in food, caulking, lubricants, kinetic sand and heat-resistant tiles. In addition to this, it serves as a critical ingredient in water-repelling coatings such as Rain-X.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Physical Form | Liquid |
Solubility Information | Miscible with carbon tetrachloride,benzene,chloroform,diethyl ether,toluene and other organic solvents. Immiscible with water and ethanol. |
Quantity | 100 g |
Chemical Name or Material | Polydimethylsiloxane (silicone) emulsion |
RUO – Research Use Only