missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Polyester Cable

Use this solid braided line with sampling equipment that weighs less than 75 lb (34 kg)

£43.35 - £52.00

Specifications

Product Type Cable
Material Polyester
Quantity 1 Ea.
View More Specs

Products 2
Product Code Brand Length (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Price Quantity & Availability  
11746088
View Documents
Cole-Parmer™
05499-36
60 m
£52.00
Each
Estimated Shipment: 04-06-2025
to see stock.
Add to basket
 
11736088
View Documents
Cole-Parmer™
05499-35
30 m
£43.35
Each
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
 
Description

Description

  • Maximum load is 110 lb (50 kg)
Specifications

Specifications

Cable
Polyester
1 Ea.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole-Parmer™ Polyester Cable

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.