Learn More
Fisherbrand™ Laser Tough-Spots™ Label Sheets
Shop All Fisherbrand Products
Click to view available options
Color:
White
Unit Size:
Each
Specifications
Specifications
| Autoclavable | Autoclavable |
| Adhesive Type | Acrylic |
| Color | White |
| Labels per Row | 12 |
| Labels per Sheet | 192 |
| No. per Pack | 3,840/PK |
| Diameter (Metric) | 9.5 mm |