Learn More
Brady™ Polyester Labels
Polyester Labels are ideal for asset tracking, barcode labelling, circuit board labelling, component and equipment labelling, data and telecommunications labelling, electrical labelling, general identification, inventory and inspection labelling.
Brand:  Brady™ 143081
 View more versions of this product
Product Code. 13521460
  • £145.00 / Each
Estimated Shipment: 27-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Abrasion-resistant, chemical-resistant, corrosion-resistant, humidity-resistant, UV-resistant, and weather-resistant.
Specifications
Specifications
Halogen Free (DIN VDE 0472 Part 815)
Polyester Labels for BBP33/i3300 Printers
White
Polyester
Labels
i3300, i5300, BBP33
-70°C
1500 Pk.
76.20 x 25.4 mm (W x H)
-94°F to +230°F
0.003 in.
28.58 mm
1 in.
3 in.
B33-18-423
Permanent Acrylic, Permanent
Gloss
Thermal Transfer
Barcode labelling, Circuit board labelling, Component and equipment labelling, General Identification, Inventory and inspection labelling
1
1500 Labels/Box
Rectangle
LabelMark, MarkWare
-70°C to +110°C
0.076 mm
25.4 x 76.2 mm
25.4 mm
76.2 mm