Learn More
Brady™ Polyester Labels for the BBP33/i3300 Printer
Our B-494 polyester label is a reliable sample identification and traceability solution.
Brand:  Brady™ 307021
Additional Details : Weight : 0.90000kg
 View more versions of this product
Product Code. 17150531
  • £205.00 / Pack of 1500
Estimated Shipment: 08-08-2024
to see stock.
Add to basket
Description
Description
  • The label will stay attached and remain legible, even when exposed to chemicals or stored in liquid nitrogen.
  • Thanks to subsurface printing, the label's colour code will not fade, smear or smudge and will continue to offer fast sample recognition long after it was printed.
Specifications
Specifications
Acrylic
Gloss
Polyester
Thermal Transfer
Non-sterile
-196°C
-320.8°F to +158°F
0.0762 mm
0.6 in.
1.625 in.
15.24 mm
Pink, White
Roll
Front
Labels
BBP™33 Label Printer; BradyPrinter i3300 Industrial Label Printer
1500 Labels/Roll
-196°C to +70°C
17.78 mm
15.24 mm
41.28 mm