Learn More
Fisherbrand™ Polyester Sealed-Edge Cleanroom Wipes
Polyester Sealed-Edge Cleanroom Wipes are absorbent and extremely clean, designed for ultra-critical environments. They contain laser cut, heat sealed-edges and constructed from double knit 100% no-run polyester.
Brand:  Fisherbrand™ 17444000
58.89 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
 View more versions of this product
Product Code. 16318317
  • £78.80 / Pack of 150
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Polyester Sealed-Edge Cleanroom Wipes are ultra-clean wipes ideal for critical cleanrooms (ISO Class 3-4 and higher). They are made from double knit 100% polyester with laser cut, heat sealed-edges. These polyester wipes are double-bagged, cleanroom packaged, and manufactured in a cleanroom environment. Wipes are manufactured and processed to meet the demands of the most critical environments. Made from the highest quality, lowest-linting substrates - free of bonding agents, adhesives or chemical additives.
Cleanroom Compatibility
  • ISO Class 3-8 (Class 1-100,000) Cleanrooms
Features
  • Manufactured from continuous run 100% polyester providing a tough, absorbent and abrasion resistant product
  • Production process guarantees extremely low particle generation and chemical extractables
  • Compatible with virtually any solvent or solution for cleaning and wiping
  • Double bagged and vacuum cleanroom packed
  • Provides lot-to-lot traceability
Specifications
Specifications
Cleanroom Wipe
22.86 cm
22.86 cm
310 mL/m2
140 gsm
White
150 Wipes/Bag, 10 Bags/Cs.
9 in.
9 in.
Polyester
Not Autoclavable
ISO Class 3-8 class 1-10 cleanrooms, Meets USP 797 and USP 800 wipe requirements
Laboratory and Cleanroom
Non-sterile