Learn More
Brady™ Polyester Thermal-Transfer Printable Labels
Polyester Thermal-Transfer Printable Labels are made of permanent polyester (B-423), which is ideal for electronic PCB and component identification, bar code labels, rating plates and solar panel identification.
Brand:  Brady™ 236713
69.74 GBP valid until 2024-12-31
Use promo code "21841" to get your promotional price.
 View more versions of this product
Product Code. 11538754
  • £77.05 / 1 roll
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Glossy white finish with permanent acrylic is thermal transfer printable
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved; CSA accepted
  • Meets requirements of a halogen-free material per DIN VDE 0472 part 815
  • Service temperature of -94 to 230° F (-70 to 110° C)
Specifications
Specifications
RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC, UL Recognized, CSA Accepted
Acrylic, Permanent
Satin
Thermal Transfer
Asset identification, rating and nameplate labels
28.4 mm
25.4W x 76.2mmH
0.017 mm
25.4 mm
76.2 mm
BPT-5-8423-2.5
White
Polyester
Thermal Transfer Printer Label
BBP11, PR200Plus, PR300Plus, PR600Plus, BP1244, BP1344, BP4000, BP4320
Rectangle
-70°C to +110°C
15.7 mm
108 mm