Learn More
Brady™ Polyester Thermal-Transfer Printable Labels
Polyester Thermal-Transfer Printable Labels are made of permanent polyester (B-423), which is ideal for electronic PCB and component identification, bar code labels, rating plates and solar panel identification.
Brand:  Brady™ 217079
 View more versions of this product
Product Code. 12882699
  • £217.00 / 1 roll
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Glossy white finish with permanent acrylic is thermal transfer printable
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved; CSA accepted
  • Meets requirements of a halogen-free material per DIN VDE 0472 part 815
  • Service temperature of -94 to 230° F (-70 to 110° C)
Specifications
Specifications
RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC, UL Recognized, CSA Accepted
Thermal Transfer Printable Labels
White
Polyester
Thermal Transfer Printer Label
BBP11, BP1244, BP1344, BP4000, BP4320
3
Rectangle
-75°C to +145°C
21.98 mm
80.58 mm
BPT-616-488
Acrylic, Permanent
Matte
Thermal Transfer
Laboratory identification
25.86 mm
3000/Pk.
22.86W x 19 mmH
0.022 mm
22.86 mm
19.05 mm