missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Polyetheretherketone Motor Valve PV-908
Motor valve PV-908, 25 Mpa, may be used as a column valve or outlet valve on ÅKTA design systems
Brand: Cytiva 18-1108-41
Code : GX
Additional Details : Weight : 1.56500kg
Description
Motor valve PV-908, 25 Mpa, may be used as a column valve or outlet valve on ÅKTA design systems. INV-907, IV-908, and PV-908 are motorised valves that are used in ÅKTAdesign platform. INV-907 is a 7-port valve (Fig 1) primarily intended for sample injection. IV-908 and PV-908 are 8-port versions for selection of columns, samples, or buffers. IV-908 can be used at pressures up to 20 bar (2 Mpa, 290 psi) and is used on the low pressure side in a system, and PV-908 and INV-907 can operate up to 250 bar (25 Mpa, 3625 psi) and are used on the high pressure side in a system. All valves are controlled via UNICORN.- A range of manual and automated valves to meet a variety of performance requirements.
- Designed for biocompatibility and years of reliable service, each may be inserted into the flow path without causing unnecessary mixing or sample dilution.
Specifications
Aqueous Solutions and Organic Solvents | |
PEEK | |
250 bar | |
8 Ports |
UniNet cable | |
PV-908 | |
Motor Valve PV-908 |