missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva PEEK Sample Loops
Resists organic solvents and salt buffers commonly used in chromatography of biomolecules. Cytiva Life Sciences™ PEEK Sample Loops are an important accessory for chromatography applications.
£171.00 - £252.00
Product Code | Brand | Diameter (Metric) Inner | Volume (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Volume (Metric) | Price | Quantity & Availability | ||||
11360032
|
Cytiva
18-1113-98 |
0.5 mm | 100 μL |
£171.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11370032
|
Cytiva
18-1114-01 |
0.75 mm | 1 mL |
£252.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11390032
|
Cytiva
18-1114-02 |
0.75 mm | 2 mL |
£194.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10852041
|
Cytiva
18-1140-53 |
1 mm | 5 mL |
£188.49
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11306971
|
Cytiva
18111399 |
0.75 mm | 500 μL |
£189.28
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Wetted parts are resistant to organic solvents
- Wetted parts are resistant to salt buffers
- Except 100 % Ethyl acetate, 100 % Hexane and 100 % Tetrahydrofuran
- Polyetheretherketone (PEEK)
Specifications
Sample Loop | |
PEEK | |
1.58 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva PEEK Sample Loops