Learn More
Cytiva Polyetheretherketone Solenoid Valve PSV-50
Excellent solvent resistance
Brand:  Cytiva 19-1994-01
Code : GE
Additional Details : Weight : 0.14800kg
Product Code. 11668883
  • £761.00 / Each
Estimated Shipment: 09-09-2024
to see stock.
Description
Description
Solenoid valves PSV-50 and the three-way valve may be operated by most fraction collectors. Both have pressure limits of 2 bar (0.2 MPa, 29 psi) and excellent solvent resistance. The PSV-50 is used for gradient formation with HiLoad Pump P-50 and flow diversion with most fraction collectors. Long life and zero dead volume make the three-way valve an excellent choice for fraction collector flow diversion in SuperFrac.
  • A range of manual and automated valves to meet a variety of performance requirements.
  • Designed for biocompatibility and years of reliable service, each may be inserted into the flow path without causing unnecessary mixing or sample dilution.
Specifications
Specifications
Ceramic, PTFE, ECTFE
PSV-50
2 bar
Solenoid Valve PSV-50