missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Polyethersulfone Membranes
Fisherbrand™ Polyethersulfone Membranes features highly asymmetrical pore structure that offers effective loading capacity and high flow rate.
£36.25 - £198.64
Specifications
Autoclavable | Yes |
---|---|
Color | White |
Material | PES |
Sterility | Non-sterile |
For Use With (Application) | Filtration of special chemical reagent, high temperature liquids, filtration of special chemical reagent, high temperature liquids, filtration of drugs |
Product Code | Brand | Pore Size | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Diameter (Metric) | Price | Quantity & Availability | |||||
11755305
|
Fisherbrand™
0442A00022 |
0.2 μm | 25 mm |
£36.25
Pack of 100 |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
11765305
|
Fisherbrand™
0442A00023 |
0.2 μm | 47 mm |
£196.56
Pack of 100 |
Estimated Shipment: 13-05-2025
Log in to see stock. |
|||||
11775305
|
Fisherbrand™
0444A00022 |
0.45 μm | 25 mm |
£83.40
Pack of 100 |
Estimated Shipment: 02-05-2025
Log in to see stock. |
|||||
11785305
|
Fisherbrand™
0444A00023 |
0.45 μm | 47 mm |
£198.64
Pack of 100 |
Estimated Shipment: 15-05-2025
Log in to see stock. |
|||||
Description
- Highly asymmetrical pore structure offers effective loading capacity and high flow rate
- Hydrophilic and made with polyethersulfone polymer
- Designed to remove particulates during general filtration
- Low protein and drug binding characteristics make it ideally suited for use in life science applications
Specifications
Yes | |
PES | |
Filtration of special chemical reagent, high temperature liquids, filtration of special chemical reagent, high temperature liquids, filtration of drugs | |
100 Pack | |
Yes |
White | |
Non-sterile | |
Hydrophilic | |
Plain Disc |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Polyethersulfone Membranes