Learn More
Velp Scientifica™ Polyethylene Bottle
Velp Scientifica™ Polyethylene Bottle is designed to fit inside an overhead mixer.
Brand:  Velp Scientifica™
Code : NEW
Additional Details : Weight : 0.18000kg
 View more versions of this product
Product Code. 15399624
  • £18.50 / Each
Estimated Shipment: 29-08-2024
to see stock.
Add to basket
Description
Description
  • For use in Rotax 6.8 Overhead Mixer
  • Constructed of Polyethylene
Specifications
Specifications
2 L
ROTAX
Bottle