missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha Packaging™ Polyethylene Clinical Waste Sack
Click to view available options
Color:
Orange
Yellow
Unit Size:
Pack of 200
Description
- Polyethylene
- For incineration
Specifications
Specifications
| Color | Orange |
| Dimensions (L x W) | 735 x 900 mm |
| For Use With (Application) | Clinical for incineration. |
| Material | Polyethylene |
| Product Type | Waste Sack |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction