Learn More
Kartell™ Polyethylene Disc Membrane
These discs are made of polyethylene and have good chemical resistance. Kartell™ Polyethylene Disc Membrane is used to separate the filter paper from the funnel plate when working with a Buchner Funnel.
Brand:  Kartell™ 0084000
8.98 GBP valid until 2025-03-30
Use promo code "24134" to get your promotional price.
 View more versions of this product
Product Code. 12677765
  • £9.45 / Pack of 10
Estimated Shipment: 10-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Useful interface between filter paper and perforated Buchner plate
  • Avoids clogging of filter holes
Specifications
Specifications
Screen Filter
Polyethylene
Interface between filter paper and perforated Buchner plate
Circle
Disc
300 μm
10/Pack
110 mm