Learn More
Masterflex™ Polyethylene Elbow Pipe Adapter
Designed for quick and easy assembly without tools
Brand: Masterflex™ 30623-12
This item is not returnable.
View return policy
Description
- Ideal for low-pressure applications
- Chemical resistant
- Available in a variety of sizes
- Economical barbed fittings easily connect fluid paths without the use of tools
- Just slip your flexible tubing or hose onto the barb to create a secure seal
Specifications
Barb x NPT(M) Connection | |
1/4 in. | |
1 21/32 in. | |
3/8 in. | |
1/2 in. | |
Ethylene Oxide | |
Elbow-shaped | |
Barbed Fitting |
Not Autoclavable | |
HDPE | |
10 Fittings | |
3/8 in. | |
3/8 in. | |
1 25/32 in. | |
Translucent |