Learn More
Brand™ Non-Sterile Polyethylene Filter Tips
Non-self-sealing filter tips from Brand™ have a PE filter that is free from chemical additives.
Brand: Brand™ 732706
46.10 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Permeability is controlled by the combination of pore size and filter length, so that no aerosols can reach the pipet shaft
- Functions with consistent reliability
- Sample can be recovered from the filter by simply actuating the pipette’s blow-out function, or by centrifugation if necessary
- Available in a wide range of capacities
Specifications
1 to 20 μL | |
Non-sterile | |
Ideal for biological samples that contain detergents such as TRITON™ X-100, SDS, Tween | |
TipBox: 5 boxes, 96 each | |
Yes |
Filtered | |
Grey | |
Filter Tip | |
480 | |
Polyethylene |