missing translation for 'onlineSavingsMsg'
Learn More
Polyethylene glycol 2,000, Thermo Scientific Chemicals Product Code.: 11469717

Polyethylene glycol 2,000, Thermo Scientific Chemicals

Product Code. 11469717
250 g
Click to view available options
Quantity:
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11469717

Brand: Thermo Scientific Alfa Aesar B22181.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Polyethylene glycol is used in the field of pharmaceuticals, cosmetics, paper, rubber and ceramics and in wood processing. It is also used in the manufacture of surfactants, dispersant resin and plastics. It acts as a dye carrier in paints and inks; soldering fluxes with good spreading property and as a softener and antistatic agent for textiles. It is also one of the main materials of ester type surface active agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym PEG 2000
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

pH 5 to 7
Melting Point 53°C to 55°C
Merck Index 14,7568
Solubility Information Soluble in water and organic solvents. Insoluble in aliphatic hydrocarbons.
Quantity 250 g
Odor Odorless
Chemical Name or Material Polyethylene glycol 2,000

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252223 Certificate TypeCertificate of Analysis Date06/09/2024 Catalog NumberB22181.30,B22181.0B,B22181.PG
Lot Number10251010 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberB22181.30,B22181.0B,B22181.PG
Lot Number10247727 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog NumberB22181.30,B22181.0B,B22181.PG
Lot Number10241359 Certificate TypeCertificate of Analysis Date17/08/2023 Catalog NumberB22181.30,B22181.0B,B22181.PG
Lot Number10247400 Certificate TypeCertificate of Analysis Date07/08/2023 Catalog NumberB22181.30,B22181.0B,B22181.PG

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Polyethylene glycol 2,000, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.