Learn More
Polyethylene glycol 400, Thermo Scientific Chemicals
Description
Polyethylene glycol is used in a variety of pharmaceutical formulations. It finds application in Hewlett-Packard designjet printers as an ink solvent and a lubricant for the print heads. It is an important raw material for the manufacture of polyuretnae, PEG salts of lauric, oleic and stearic acids and latex.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
pH | 5 to 7 |
Melting Point | 4°C to 8°C |
Linear Formula | H(OCH2CH2)nOH |
Density | 1.126 |
Merck Index | 14,7568 |
Solubility Information | Miscible with water,acetone,alcohols,benzene,glycerin,glycols and aromatic hydrocarbons. Slightly miscible with aliphatic hydrocarbons. |
Quantity | 250 g |
Flash Point | 240°C (464°F) |
Odor | Odorless |
Refractive Index | 1.466 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10249059 | Certificate of Analysis | 12/08/2024 | B21992.0B,B21992.30 |
10240380 | Certificate of Analysis | 29/04/2022 | B21992.0B,B21992.30 |
10238470 | Certificate of Analysis | 12/04/2022 | B21992.0B,B21992.30 |
10213733 | Certificate of Analysis | 23/11/2021 | B21992.0B,B21992.30 |
10180125 | Certificate of Analysis | 23/11/2021 | B21992.0B,B21992.30 |
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
58Hours
16Minutes
5Seconds
32New Offer Save Up to 70%
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.