missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene Glycol 8000 (PEG), Fisher BioReagents™
50.30 GBP valid until 2024-12-31
Use promo code "23286" to get your promotional price.
Use promo code "23286" to get your promotional price.
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass Bottle
Poly Bottle
Description
Polyethylene Glycol 8000, HO-[CH2CH2O]n-H, CAS Number-25322-68-3, Polyethylene Oxide, Carbowax, PEG 8000, 1kg, Gray, White, 7000 to 9000, CAUTION!
PEG 8000 is suitable as a medium for the fusion of mammalian cells. It is also used to precipitate bacteriophage from lysed cell supernatants.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | White |
pH | 5 to 7 |
Physical Form | Solid |
Melting Point | 55°C |
Linear Formula | H(OCH2CH2)nOH |
Merck Index | 15, 7688 |
Quantity | 1 kg |
Formula Weight | 7000 to 9000 |
Identification | Pass Test |
Packaging | Poly Bottle |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'