missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene Glycol Antibody (RM105), Biotin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-18537
Additional Details : Weight : 0.00970kg
Description
Polyethylene Glycol Monoclonal antibody specifically detects Polyethylene Glycol in All samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
Polyethylene Glycol | |
Monoclonal | |
1 mg/ml | |
Western Blot 0.1 to 1 μg/mL, ELISA : 0.02 to 0.5 μg/mL, Immunohistochemistry 0.5 to 2 μg/mL | |
1,2-Dihydroxyethane, 1,2-ethanediol, 107-21-1, 2-hydroxyethanol, Ethane-1,2-diol, glycol, Glycol alcohol, monoethylene glycol | |
KLH conjugated Polyethylene Glycol (PEG) with terminal methoxy group | |
50 μg | |
Primary | |
Store at -20°C. | |
IgG |
Western Blot, ELISA, Immunohistochemistry | |
RM105 | |
Biotin | |
50% Glycerol, PBS, 1% BSA | |
Rabbit | |
Protein A purified | |
RUO | |
All species | |
Purified |