missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol di-p-toluenesulfonate, M.W. 3,400, Thermo Scientific™

Product Code. 15407355
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula CH3C6H4SO2(OCH2CH2)nOSO2C6H4CH3
Synonym PEG-di-p-tosylate

Specifications

Color White
Physical Form Granular Solid
Melting Point 43°C to 47°C
Quantity 5 g
Chemical Name or Material Polyethylene glycol di-p-toluenesulfonate, M.W. 3,400
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'