missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene glycol di-p-toluenesulfonate, M.W. 3,400, Thermo Scientific™
£34.40 - £152.00
Chemical Identifiers
Molecular Formula | CH3C6H4SO2(OCH2CH2)nOSO2C6H4CH3 |
---|---|
Synonym | PEG-di-p-tosylate |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
CH3C6H4SO2(OCH2CH2)nOSO2C6H4CH3 | |
PEG-di-p-tosylate |
Specifications
White | |
43°C to 47°C | |
PEG-di-p-tosylate |
Granular Solid | |
CH3C6H4SO2(OCH2CH2)nOSO2C6H4CH3 | |
Polyethylene glycol di-p-toluenesulfonate, M.W. 3,400 |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures