missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene glycol diacrylate, M.W. 3,400, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
Polyethylene glycol diacrylate is used in dental compounds, offset inks, varnishes and flexible coatings. It widely used in various biomedical applications such as hydrogel development for drug delivery, tissue engineering, wound healing and electrospinning processes for bioapplication due to its non-toxicity and cytocompatiblity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | White |
Physical Form | Granular Solid |
Solubility Information | Miscible with water. Soluble in THF,Chloroform. |
Quantity | 1 g |
Sensitivity | Light sensitive |
Chemical Name or Material | Polyethylene glycol diacrylate, M.W. 3,400 |
Safety and Handling
Product Identifier
- Polyethylene glycol diacrylate, M.W. 3,400
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- MIXTURE LIST-Contains : .alpha.-(1-oxo-2-propenyl)-.omega.-[(1-oxo2-propenyl)oxy]-Poly(oxy-1,2-ethanediyl)
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'