missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene glycol diacrylate, M.W. 3,400, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
Polyethylene glycol diacrylate is used in dental compounds, offset inks, varnishes and flexible coatings. It widely used in various biomedical applications such as hydrogel development for drug delivery, tissue engineering, wound healing and electrospinning processes for bioapplication due to its non-toxicity and cytocompatiblity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | White |
Physical Form | Granular Solid |
Solubility Information | Miscible with water. Soluble in THF,Chloroform. |
Quantity | 250 mg |
Sensitivity | Light sensitive |
Chemical Name or Material | Polyethylene glycol diacrylate, M.W. 3,400 |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'