missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene glycol-lithium acetate soln., Thermo Scientific™
Click to view available options
Quantity:
250 mL
500 mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Colorless |
Quantity | 250 mL |
Physical Form | Liquid |
Chemical Name or Material | Polyethylene glycol-lithium acetate soln. |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'