missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol methyl ether methacrylate, M.W. 13,000, Thermo Scientific Chemicals

Product Code. 15467235
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Polyethylene glycol methyl ether methacrylate, is used as Emulsion polymers, cosmetics, construction, emulsifiers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26915-72-0
Molecular Formula C13H24O6
Molecular Weight (g/mol) 276.329
MDL Number MFCD00241432
InChI Key KRCGBOKYIUDIFY-UHFFFAOYSA-N
Synonym 2-methoxyethyl methacrylate, beta-methoxyethyl methacrylate, methacrylic acid, 2-methoxyethyl ester, 2-propenoic acid, 2-methyl-, 2-methoxyethyl ester, poly ethylene glycol methyl ether methacrylate, pegmem, peg-mem, acmc-209oay, methoxyethyl methacrylate, 2-methoxyethylmethacrylate
PubChem CID 93675
IUPAC Name 2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCOCCOCCOCCOC

Specifications

Color White
Physical Form Granular Solid
Solubility Information Soluble in water.
Quantity 250 mg
Chemical Name or Material Polyethylene glycol methyl ether methacrylate, M.W. 13,000

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'