Learn More
Polyethylene glycol methyl ether methacrylate, M.W. 5,000, Thermo Scientific Chemicals
£28.59 - £325.62
Chemical Identifiers
CAS | 26915-72-0 |
---|---|
Molecular Formula | C13H24O6 |
Molecular Weight (g/mol) | 276.329 |
MDL Number | MFCD00241432 |
InChI Key | KRCGBOKYIUDIFY-UHFFFAOYSA-N |
Synonym | 2-methoxyethyl methacrylate, beta-methoxyethyl methacrylate, methacrylic acid, 2-methoxyethyl ester, 2-propenoic acid, 2-methyl-, 2-methoxyethyl ester, poly ethylene glycol methyl ether methacrylate, pegmem, peg-mem, acmc-209oay, methoxyethyl methacrylate, 2-methoxyethylmethacrylate |
PubChem CID | 93675 |
IUPAC Name | 2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethyl 2-methylprop-2-enoate |
SMILES | CC(=C)C(=O)OCCOCCOCCOCCOC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15447335
|
Thermo Scientific Alfa Aesar
046838.MD |
250 mg |
£28.59
250mg |
Estimated Shipment: 03-12-2024
Log in to see stock. |
|||||
15427335
|
Thermo Scientific Alfa Aesar
046838.03 |
1 g |
£78.59
1g |
In Stock
Log in to see stock. |
|||||
15437335
|
Thermo Scientific Alfa Aesar
046838.06 |
5 g |
£325.62
5g |
In Stock
Log in to see stock. |
|||||
Description
Polyethylene glycol methyl ether methacrylate, is used as Emulsion polymers, cosmetics, Construction, emulsifiers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
26915-72-0 | |
276.329 | |
KRCGBOKYIUDIFY-UHFFFAOYSA-N | |
93675 | |
CC(=C)C(=O)OCCOCCOCCOCCOC |
C13H24O6 | |
MFCD00241432 | |
2-methoxyethyl methacrylate, beta-methoxyethyl methacrylate, methacrylic acid, 2-methoxyethyl ester, 2-propenoic acid, 2-methyl-, 2-methoxyethyl ester, poly ethylene glycol methyl ether methacrylate, pegmem, peg-mem, acmc-209oay, methoxyethyl methacrylate, 2-methoxyethylmethacrylate | |
2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethyl 2-methylprop-2-enoate |
Specifications
White | |
Granular Solid | |
MFCD00241432 | |
It is soluble in water. | |
CC(=C)C(=O)OCCOCCOCCOCCOC | |
276.329 | |
Polyethylene glycol methyl ether methacrylate, M.W. 5,000 |
26915-72-0 | |
C13H24O6 | |
2-methoxyethyl methacrylate, beta-methoxyethyl methacrylate, methacrylic acid, 2-methoxyethyl ester, 2-propenoic acid, 2-methyl-, 2-methoxyethyl ester, poly ethylene glycol methyl ether methacrylate, pegmem, peg-mem, acmc-209oay, methoxyethyl methacrylate, 2-methoxyethylmethacrylate | |
KRCGBOKYIUDIFY-UHFFFAOYSA-N | |
2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethyl 2-methylprop-2-enoate | |
93675 |
Safety and Handling
GHS H Statement
H318-H317-H335-H315
Causes serious eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P333+P313-P362-P501c
H315-H317-H318-H335
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only