missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol monomethyl ether methanesulfonate, M.W. 5,000, Thermo Scientific™

Product Code. 15487215
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 175172-61-9
Molecular Formula C4H10O4S
Molecular Weight (g/mol) 154.18
MDL Number MFCD03100948
InChI Key BCKAHDGFNHDQST-UHFFFAOYSA-N
Synonym methanesulfonic acid 2-methoxyethyl ester, ethanol, 2-methoxy-, methanesulfonate, 2-methoxyethanol methanesulfonate, 2-methoxyethyl mesylate, 2-methoxyethylmethanesulfonate, acmc-1c990, 2-methoxyethyl methane sulfonate, methanesulfonic acid2-methoxyethyl ester, ethanol, 2-methoxy-, 1-methanesulfonate, methanesulfonic acid 2-methoxy-ethyl ester
PubChem CID 146293
IUPAC Name 2-methoxyethyl methanesulfonate
SMILES COCCOS(=O)(=O)C

Spezifikation

Color White
Physical Form Granular Solid
Melting Point 58°C to 61°C
Quantity 1 g
Chemical Name or Material Polyethylene glycol monomethyl ether methanesulfonate, M.W. 5,000
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'