Learn More
Kartell™ Plastilab™ Graduated Wide Mouth Wash Bottles
Wide mouth design aids easy and safe filling of contents.
Brand:  Kartell™ 0163810
Product Code. 10760155
  • ONLINE EXCLUSIVE PRICE
    £41.45
    £36.66 / Pack of 10
    Save £4.79
    11% Off
Only null left
Estimated Shipment: 23-01-2026
Add to basket
Description
Description
Wide mouth design aids easy and safe filling of contents.
  • Made from polyethylene
  • Color-coded caps for easy identification
  • Ingenious cap designed to avoid involuntary dripping
  • Suitable for storing food stuff
Specifications
Specifications
500 mL
Red
Polyethylene
Red
Yes
Wide
16.9 oz.
Color coded cap
GL45
100mL
Yes