missing translation for 'onlineSavingsMsg'
Learn More

Gosselin™ Polyethylene Homogenizer Blender Bags

View contents easily through clear polyethylene. Gosselin™ Polyethylene Homogenizer Blender Bags include white writing strip along one edge for content identification.

Brand:  Gosselin BBAG-03

 View more versions of this product

Product Code. 11857403

  • £824.73 / Pack of 1500
Estimated Shipment: 05-08-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Sterile by irradiation with peel away sterilization certificate
  • Available with full or side filter
  • Includes 50mm white writing strip along one long edge
  • 70μm thickness
TRUSTED_SUSTAINABILITY
Specifications

Specifications

400 mL
Ideal for homogenous suspension, suspension sampling with pipets, dairy and industries
Polyethylene
70 μm
1500/Pk.
190 mm
Natural
Side filter
Sterile
20 bags of 25
300 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gosselin™ Polyethylene Homogenizer Blender Bags > Side filter; Packaged in 20 bags of 25

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.