missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Polyethylene Hose Couplings

Product Code. 10161782 Shop All Bürkle Products
Click to view available options
Diameter (Metric):
11 to 14 mm
13 to 16 mm
3 to 5 mm
5 to 7 mm
7 to 10 mm
9 to 12 mm
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 10161782

Brand: Buerkle™ 87041214

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Attachable, conical hose couplings for pressureless operation.

  • Manufactured from polyethylene
  • Provided 10 per pack
  • Available for a range of hose diameters
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EU Foodstuffs, RoHS
Diameter (Metric) 11 to 14 mm
Autoclavable Autoclavable
Material Polyethylene
For Use With 11 to 14 mm Hose
For Use With (Application) Pressureless Operation
Packaging Quantity 10/Pk.
Shape Conical
Product Type Hose Coupling
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.