Learn More
Kartell™ Polyethylene Integral Eye Wash Bottle
Equipped with an eyecup and specially designed snap-on cap for fast and convenient eye washing to remove foreign objects quickly and prevent serious damage to vision. Kartell™ Polyethylene Integral Eye Wash Bottles can be filled with distilled water or any other medicated liquid intended for ophthalmic use.
Brand: Kartell™ 0038300
126.35 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
Description
Equipped with an eyecup and specially designed snap-on cap for fast and convenient eye washing to remove foreign objects quickly and prevent serious damage to vision. Kartell™ Polyethylene Integral Eye Wash Bottles can be filled with distilled water or any other medicated liquid intended for ophthalmic use.
- Automatic valve incorporated in the head to admmit air and prevent contaminated eyewash from flowing back into bottle
- Perforated plate in eyecup breaks up the stream and provides a uniform spray to the eye
- Includes a tube to drain the contaminated eyewash
- Suitable for foodstuff
Specifications
500 mL | |
Polyethylene | |
73 mm |
213 mm | |
Integral Eye Wash Bottle |