Learn More
Kartell™ Centrifuge Microtubes
Designed to withstand a centrifuge force up to RCF 11,000. Kartell™ Centrifuge Microtubes are ideal for sample analysis.
Brand: Kartell™ 0129900
9.60 GBP valid until 2024-07-31
Use promo code "21640" to get your promotional price.
Code : 02
Additional Details : Weight : 0.54000kg
Description
Designed to withstand a centrifuge force up to RCF 11,000. Kartell™ Centrifuge Microtubes are ideal for sample analysis.
- Available with cap
- Manufactured from polypropylene or polyethylene
- Autoclavable
- CE marked
- IVD approved