missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene oxide, M.W. 300,000, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Polyethylene oxide, M.W. 300,000 is used to impart viscosity to and modify flow of aqueous solutions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | White |
pH | 5 to 7 |
Physical Form | Powder |
Melting Point | 65°C |
Density | 1.21 g/mL |
Solubility Information | Soluble in acetone,alcohol,chloroform,toluene and dichloromethane. |
Quantity | 25 g |
Odor | Odorless |
Refractive Index | 1.4539 |
Chemical Name or Material | Polyethylene oxide, M.W. 300,000 |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'