missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Polyethylene PCR Microplate Sealing Tape
Brand: Corning™ 6524AL
Code : 66
Additional Details : Weight : 0.18000kg
Description
Thermowell™ GOLD PCR consist of five product groups: 0.2mL and 0.5mL individual PCR tubes, 1 x 8 0.2mL tube strips and caps (including optically clear flat cap strips for RT-PCR), 96 well polypropylene PCR plates, 384 well polypropylene PCR microplates, and sealers for PCR reaction vessels. Each group features a variety of formats to ensure compatibility with thermocycling and sequencing instrumentation, application requirements and user preferences.- Tubes and plates feature ultra thin and uniform wall thickness
- Longer cap flanges on tubes and strips
- Full skirt, half skirt and elevated skirt formats for 96 well plates
- Optically clear 1 x 8 flat cap strips and tape
- Certified RNase/DNase free For PCR* and quantitative real-time PCR
Specifications
Polyethylene | |
Autoclavable | |
100 Each | |
yes |
Non-sterile | |
100 Each | |
PCR Microplate Sealing Tape |