Learn More
Polyethylene QC Standard Blank, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep CRM-PEBLK
Code : NEW
Additional Details : CAS Number : 9002-88-4 Weight : 0.05000kg
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
Bis(2-Ethylhexyl)phthalate | 117-81-7 | 0μg/g |
Butylbenzyl phthalate | 85-68-7 | 0μg/g |
Diethyl phthalate | 84-66-2 | 0μg/g |
Diisodecyl phthalate | 26761-40-0 | 0μg/g |
Diisononyl phthalate | 28553-12-0 | 0μg/g |
Dimethyl phthalate | 131-11-3 | 0μg/g |
Di-n-butyl phthalate | 84-74-2 | 0μg/g |
Di-n-octyl phthalate | 117-84-0 | 0μg/g |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Polyethylene QC Standard Blank | |
5 g | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
2mL amber prescored glass ampule | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. | |
Polyethylene |
Product intended for laboratory use only