missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polygon Magnetic Stirring Bars without Pivot Ring

These stirbars have polygon multifaceted surfaces that add turbulence relative to similar smooth size cylindrical bars

Brand:  Fisherbrand™ F37120-0020

Product Code. 15588682

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Description

Description

  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
  • Interrupted profile provides greater surface area and added turbulence when compared to the smooth surface of cylindrical bars
  • Pivot ring aids in reducing friction and chattering
  • Autoclavable
  • Manufactured in registered ISO 9001:2008 facilities, as verified by SGS certification
Specifications

Specifications

Autoclavable
White
0.3125 in.
8 mm
Alnico Magnet
ISO 9001:2015
Stirring Bar
0.75 in.
20 mm
Polygon
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers