Learn More
Macherey-Nagel™ Polygoprep™ 300 to 20 C18
Brand:  Macherey-Nagel™ 711025.100
 View more versions of this product
Product Code. 10747462
  • £363.00 / 100g
Estimated Shipment: 24-12-2024
to see stock.
Explore more special offers
Add to basket
Specifications
15 to 25 μm
POLYGOPREP 300 to 20
0.1 kg
Polygoprep
300 Å
2 to 8
POLYGOPREP C18
Liquid Chromatography