missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Polygoprep™ 60 to 12 NH2

£214.00 - £1575.00

Specifications

Particle Size 10 to 15 μm
Pore Size 60 Å
Stationary Phase POLYGOPREP 60 to 12
Technique Liquid Chromatography
pH 2 to 8
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10462032
View Documents
Macherey-Nagel™
711012.100
0.1 kg
£214.00
100g
Estimated Shipment: 14-03-2025
to see stock.
Add to basket
 
10104551
View Documents
Macherey-Nagel™
711012.1000
1 kg
£1575.00
1000g
Estimated Shipment: 14-03-2025
to see stock.
Add to basket
 
Description

Description

Specifications

Specifications

10 to 15 μm
POLYGOPREP 60 to 12
2 to 8
350 m2/g
Silica
60 Å
Liquid Chromatography
POLYGOPREP NH2
Polygoprep
Liquid Chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Macherey-Nagel™ Polygoprep™ 60 to 12 NH2

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.